Please login to your online account to display your discounted pricing

1,8-Diaminonaphthalene, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC371180050

 View more versions of this product

Catalog No. AC371180050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Color Grayish-red to gray or dark brown
Formula Weight 158.2
Physical Form Chunks or Crystalline Mass
Quantity 5g
Beilstein 13, 205
Merck Index 15, 6457
Melting Point 61° to 66°C
CAS Max % 100.0
CAS Min % 96.0
Infrared Spectrum Authentic
MDL Number MFCD00004033
Name Note 97%
Linear Formula C10H6(NH)
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Synonym 1, 8-Naphthalenediamine
Packaging Glass bottle
Molecular Formula C10H10N2
Chemical Name or Material 1, 8-Diaminonaphthalene
Assay Percent Range 97%
CAS 479-27-6
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Suspected of causing cancer.
May cause respiratory irritation.
Harmful if swallowed.

Specifications

Chemical Name or Material 1, 8-Diaminonaphthalene
Name Note 97%
CAS 479-27-6
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10H10N2
Linear Formula C10H6(NH)
Formula Weight 158.2
MDL Number MFCD00004033
Synonym 1, 8-Naphthalenediamine
Accessories

GHS P Statement:
Wear eye protection/face protection.
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Suspected of causing cancer.
May cause respiratory irritation.
Harmful if swallowed.

SDS & Safety Documents