Please login to your online account to display your discounted pricing

1,8-Diamino-3,6-dioxaoctane, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1, 8-Diamino-3, 6-dioxaoctane
CAS 929-59-9
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C6H16N2O2
Linear Formula NH2CH2CH2OCH2CH2OCH2CH2NH2
Formula Weight 148.21
MDL Number MFCD00040474
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC275850050

  SDS
thermo fisher scientific
275850050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 148.21
Quantity 5g
Density 1.0360g/mL
CAS Max % 100.0
Chemical Name or Material 1, 8-Diamino-3, 6-dioxaoctane
Packaging Glass bottle
Molecular Formula C6H16N2O2
Linear Formula NH2CH2CH2OCH2CH2OCH2CH2NH2
CAS 929-59-9
Assay Percent Range 98%
MDL Number MFCD00040474
CAS Min % 97.5
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.