Please login to your online account to display your discounted pricing

1,8-Diamino-3,6-dioxaoctane, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  275850050

 View more versions of this product

Catalog No. AC275850050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 148.21
Quantity 5g
Density 1.0360g/mL
CAS Max % 100.0
Linear Formula NH2CH2CH2OCH2CH2OCH2CH2NH2
Molecular Formula C6H16N2O2
Chemical Name or Material 1, 8-Diamino-3, 6-dioxaoctane
Packaging Glass bottle
CAS 929-59-9
Assay Percent Range 98%
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 97.5
MDL Number MFCD00040474

Specifications

Chemical Name or Material 1, 8-Diamino-3, 6-dioxaoctane
CAS 929-59-9
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C6H16N2O2
Linear Formula NH2CH2CH2OCH2CH2OCH2CH2NH2
Formula Weight 148.21
MDL Number MFCD00040474
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.

SDS & Safety Documents