missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,8-Cineole, 99%

Manufacturer:  Alfa Aesar™ A12269AP

 View more versions of this product

Catalog No. AAA12269AP


 

Description

Description

Applications
Eucalyptol is an ingredient in many brands of mouthwash and cough suppressant. It controls airway mucus hypersecretion and asthma via anti-inflammatory cytokine inhibition. Eucalyptol is an effective treatment for nonpurulent rhinosinusitis. Eucalyptol reduces inflammation and pain when applied topically. It kills leukaemia cells in vitro.

Solubility
Soluble in water(3500 mg/L (at 21°C). Miscible with ether, alcohol, chloroform, glacial acetic acid, oils. Soluble in ethanol, ethyl ether; slightly soluble in carbon tetrachloride.

Notes
Store this chemical under refrigerated temperatures and away from mineral acids and bases. Store in a tightly closed container. Store in a dry, well-ventilated area.
Specifications

Specifications

1,8-Cineole
470-82-6
99%
MFCD00167977
105109
1.0°C
500mL
176°C to 177°C
2,2,4-trimethyl-3-oxabicyclo[2.2.2]octane
2758
Liquid
49°C (120°F)
99%
C10H18O
UN3271
14,3895
Eucalyptol; 1,3,3-Trimethyl-2-oxabicyclo[2.2.2]octane
WEEGYLXZBRQIMU-UHFFFAOYSA-N
CC1(C2CCC(O1)(CC2)C)C
154.253
154.24

Chemical Identifiers

470-82-6
154.253
WEEGYLXZBRQIMU-UHFFFAOYSA-N
2758
C10H18O
MFCD00167977
Eucalyptol; 1,3,3-Trimethyl-2-oxabicyclo[2.2.2]octane
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.

RTECSNumber : OS9275000

TSCA : TSCA

SDS Documents