Learn More
1,8-Bis(dimethylamino)naphthalene, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0031309
Description
1,8-Bis(dimethylamino)naphthalene is used in the synthesis of C-substituted t-BuNH-8,9-R,R?-nido-7,8,9-C3B8H9 (R,R? = H,H; MeH; Me,Me; Ph,H and Ph,Ph) tricarbollide compounds. It was also used in the preparation of saturated fluoroalkyl(hydrido) complexes of Iridium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,8-Bis(dimethylamino)naphthalene is used in the synthesis of C-substituted t-BuNH-8,9-R,R′-nido-7,8,9-C3B8H9 (R,R′ = H,H; MeH; Me,Me; Ph,H and Ph,Ph) tricarbollide compounds. It was also used in the preparation of saturated fluoroalkyl(hydrido) complexes of Iridium.
Solubility
Soluble in organic solvents.
Notes
Air & Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
1,8-Bis(dimethylamino)naphthalene | |
47°C to 51°C | |
>110°C (230°F) | |
MFCD00003920 | |
UN3263 | |
Air and light sensitive | |
Soluble in organic solvents. | |
CN(C)C1=CC=CC2=C1C(=CC=C2)N(C)C | |
214.312 | |
214.31 |
20734-58-1 | |
144°C to 145°C (7 mmHg) | |
C14H18N2 | |
10 g | |
396782 | |
1,8-bis dimethylamino naphthalene, n1,n1,n8,n8-tetramethylnaphthalene-1,8-diamine, proton sponge, 1,8-bis dimethylamino naphtalene, n,n,n',n'-tetramethyl-1,8-naphthalenediamine, proton-sponge, 1,8-naphthalenediamine, n,n,n',n'-tetramethyl, unii-6s79d2p9h8, 1,8-bis-dimethylamino naphthalene, n,n,n',n'-tetramethylnaphthalene-1,8-diamine | |
GJFNRSDCSTVPCJ-UHFFFAOYSA-N | |
1-N,1-N,8-N,8-N-tetramethylnaphthalene-1,8-diamine | |
88675 | |
≥98% |
Chemical Identifiers
20734-58-1 | |
214.312 | |
GJFNRSDCSTVPCJ-UHFFFAOYSA-N | |
88675 | |
CN(C)C1=CC=CC2=C1C(=CC=C2)N(C)C |
C14H18N2 | |
MFCD00003920 | |
1,8-bis dimethylamino naphthalene, n1,n1,n8,n8-tetramethylnaphthalene-1,8-diamine, proton sponge, 1,8-bis dimethylamino naphtalene, n,n,n',n'-tetramethyl-1,8-naphthalenediamine, proton-sponge, 1,8-naphthalenediamine, n,n,n',n'-tetramethyl, unii-6s79d2p9h8, 1,8-bis-dimethylamino naphthalene, n,n,n',n'-tetramethylnaphthalene-1,8-diamine | |
1-N,1-N,8-N,8-N-tetramethylnaphthalene-1,8-diamine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, BASIC, ORGANIC, N.O.S.
EINECSNumber : 244-001-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only