missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,8-Bis(2-pyridyl)-3,6-dithiaoctane, TCI America™

$381.95 - $1,185.58

Chemical Identifiers

CAS 64691-70-9
Molecular Formula C16H20N2S2
Molecular Weight (g/mol) 304.47
MDL Number MFCD00006366
InChI Key YDPDVMHQFYTZDI-UHFFFAOYSA-N
Synonym S,S′-Bis[2-(2-pyridyl)ethyl]-1,2-ethanedithiol, 2,2′-(3,6-Dithiaoctamethylene)pyridine
PubChem CID 300754
IUPAC Name 2-[2-[2-(2-pyridin-2-ylethylsulfanyl)ethylsulfanyl]ethyl]pyridine
SMILES C1=CC=NC(=C1)CCSCCSCCC2=CC=CC=N2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B12291G View Documents TCI America
B12291G
1g Each for $381.95
 
B12295G View Documents TCI America
B12295G
5g Each for $1,185.58
 
Specifications

Chemical Identifiers

64691-70-9
304.47
YDPDVMHQFYTZDI-UHFFFAOYSA-N
300754
C1=CC=NC(=C1)CCSCCSCCC2=CC=CC=N2
C16H20N2S2
MFCD00006366
S,S′-Bis[2-(2-pyridyl)ethyl]-1,2-ethanedithiol, 2,2′-(3,6-Dithiaoctamethylene)pyridine
2-[2-[2-(2-pyridin-2-ylethylsulfanyl)ethylsulfanyl]ethyl]pyridine

Specifications

Undesignated
1g
C16H20N2S2
S,S′C-Bis[2-(2-pyridyl)ethyl]-1,2-ethanedithiol, 2,2′C-(3,6-Dithiaoctamethylene)pyridine
C1=CC=NC(=C1)CCSCCSCCC2=CC=CC=N2
304.47
304.47
≥98.0% (GC,T)
51°C
64691-70-9
MFCD00006366
YDPDVMHQFYTZDI-UHFFFAOYSA-N
2-[2-[2-(2-pyridin-2-ylethylsulfanyl)ethylsulfanyl]ethyl]pyridine
300754
Crystal-Powder at 20°C
1,8-Bis(2-pyridyl)-3,6-dithiaoctane
Safety and Handling

Safety and Handling

TSCA : No