Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,8,9-Trihydroxyanthracene, 97%

$49.11 - $175.47

Chemical Identifiers

Chemical Name or Material 1,8,9-Trihydroxyanthracene
CAS 1143-38-0
Molecular Formula C14H10O3
Formula Weight 226.23
MDL Number MFCD00001250
Beilstein 2054360
Merck Index 14,684
Synonym Dithranol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2030303

 
alfa aesar™
B2030303
1g Each for $49.11

AAB2030306

 
alfa aesar™
B2030306
5g Each for $175.47
Description & Specifications

Specifications

Formula Weight 226.23
Chemical Name or Material 1,8,9-Trihydroxyanthracene
Assay 97%
Molecular Formula C14H10O3
MDL Number MFCD00001250
CAS 1143-38-0
Beilstein 2054360
Merck Index 14,684
Synonym Dithranol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number CB1225000
Melting Point 177° to 181°C
Quantity 1g

Applications
Dithranol was used as a matrix in MALDI TOF mass spectrometry measurements of sol-gel films for DNA microarray applications. It was also used as matrix in matrix-assisted laser desorption/ionization-mass spectrometry (MALDI-MS) and liquid secondary ion-tandem mass spectrometry (LSI-MS/MS) for the analysis of synthetic polymers to generate values for the average mass and the mass of the end groups. It was also used in MALDI-MS for direct detection of condensed tannins in bark. It is successfully used for the treatment of psoriasis.

Solubility
Insoluble in water. Soluble in acetic acid, chloroform (20 mg/mL), acetone, benzene, solutions of alkali hydroxides, fixed oil. Slightly soluble in 95% ethanol, alcohol, ether, and glacial acetic acid.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.