Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,7-Phenanthroline

Chemical Identifiers

Chemical Name or Material 1,7-Phenanthroline
CAS 230-46-6
Formula Weight 180.21
MDL Number MFCD00004971
UN Number UN2811
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3090903

 
alfa aesar™
3090903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3090906

 
alfa aesar™
3090906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA30909MD

 
alfa aesar™
30909MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 180.21
Chemical Name or Material 1,7-Phenanthroline
Odor Odorless
Physical Form Crystalline
MDL Number MFCD00004971
CAS 230-46-6
UN Number UN2811
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H341-H302-H312
Toxic if inhaled.
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful in contact with skin.
Melting Point 77° to 81°C
Health Hazard 3 GHS P Statement
P261-P280-P281-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECS Number SF8290000
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P261-P280-P281-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H341-H302-H312
Toxic if inhaled.
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful in contact with skin.