missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,7-Octadiyne 98.0+%, TCI America™

Manufacturer:  TCI America O014725ML

 View more versions of this product

Catalog No. O014725ML



Specifications

Specifications

1,7-Octadiyne
Yellow
871-84-1
MFCD00008580
1,7 octadiyne
C#CCCCCC#C
106.168
106.17
≥98.0% (GC)
135°C
25mL
C8H10
3295
DSOJWVLXZNRKCS-UHFFFAOYSA-N
octa-1,7-diyne
70099
Clear Liquid at 20°C

Chemical Identifiers

871-84-1
106.168
DSOJWVLXZNRKCS-UHFFFAOYSA-N
70099
C#CCCCCC#C
C8H10
MFCD00008580
1,7 octadiyne
octa-1,7-diyne
Safety and Handling

Safety and Handling

TSCA : Yes

Documents