Please login to your online account to display your discounted pricing

1,7-Octadiene, 98.50%, ACROS Organics™


Quantity 5mL

Manufacturer: thermo fisher scientific  129350050

 View more versions of this product

Catalog No. AC129350050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.0%
Formula Weight 110.2
Physical Form Liquid
Quantity 5mL
Color Clear colorless to slightly yellow
Density 0.74g/mL
UN Number 2309
CAS Min % 98.0
CAS Max % 100.0
Linear Formula H2CCH(CH2)4CH=CH2
Assay Percent Range 98.50%
Chemical Name or Material 1, 7-Octadiene
CAS 3710-30-3
Molecular Formula C8H14
Health Hazard 2 GHS H Statement:
Highly flammable liquid and vapor.
MDL Number MFCD00008668
Refractive Index 1.4210 to 1.4230 (20°C, 589nm)
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge

Specifications

Chemical Name or Material 1, 7-Octadiene
CAS 3710-30-3
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98.50%
Molecular Formula C8H14
Linear Formula H2CCH(CH2)4CH=CH2
Formula Weight 110.2
MDL Number MFCD00008668
Accessories

GHS Signal Word: Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge

GHS H Statement:
Highly flammable liquid and vapor.

SDS & Safety Documents