Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,7-Heptanedioic acid, 98%

Chemical Identifiers

Chemical Name or Material 1,7-Heptanedioic acid
CAS 111-16-0
Formula Weight 160.16g/mol
MDL Number MFCD00004425
Merck Index 14,7431
Synonym Pimelic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3952309

 
alfa aesar™
39523-09
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3952318

 
alfa aesar™
39523-18
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 103° to 105°C
Quantity 10g
Odor Odorless
Chemical Name or Material 1,7-Heptanedioic acid
CAS 111-16-0
Physical Form Powder
Assay 98%
Formula Weight 160.16g/mol
MDL Number MFCD00004425
Merck Index 14,7431
Synonym Pimelic acid
Solubility Information Soluble in alcohol, ether
TSCA TSCA