Please login to your online account to display your discounted pricing

1,7-Dioxaspiro(5.5)undecane, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC356460050

 View more versions of this product

Catalog No. AC356460050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Formula Weight 156.23
Physical Form Liquid
Quantity 5g
Color Clear colorless
Density 1.0200g/mL
CAS Max % 100.0
CAS Min % 96.0
Infrared Spectrum Authentic
Chemical Name or Material 1, 7-Dioxaspiro(5.5)undecane
Molecular Formula C9 H16 O2
Refractive Index 1.4620 to 1.4640 (20°C, 589nm)
Packaging Glass bottle
CAS 180-84-7
MDL Number MFCD00011578
Name Note 97%
Assay Percent Range 97%

Specifications

Chemical Name or Material 1, 7-Dioxaspiro(5.5)undecane
Name Note 97%
CAS 180-84-7
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9 H16 O2
Formula Weight 156.23
MDL Number MFCD00011578