Please login to your online account to display your discounted pricing

1,7-Dioxaspiro(5.5)undecane, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1, 7-Dioxaspiro(5.5)undecane
Name Note 97%
CAS 180-84-7
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9 H16 O2
Formula Weight 156.23
MDL Number MFCD00011578
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC356460050

  SDS
thermo fisher scientific
AC356460050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 156.23
Quantity 5g
Color Clear colorless
Density 1.0200g/mL
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 1, 7-Dioxaspiro(5.5)undecane
Packaging Glass bottle
Refractive Index 1.4620 to 1.4640 (20°C, 589nm)
Molecular Formula C9 H16 O2
MDL Number MFCD00011578
CAS 180-84-7
Infrared Spectrum Authentic
Name Note 97%
Assay Percent Range 97%