Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,7-Dimethylnaphthalene, 97%

$184.72 - $184.72

Chemical Identifiers

Chemical Name or Material 1,7-Dimethylnaphthalene
CAS 575-37-1
Molecular Formula C12H12
Formula Weight 156.23
MDL Number MFCD00060884
Beilstein 2039377
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA18030ME

 
alfa aesar™
A18030ME
500mg Each for $184.72
Description & Specifications

Specifications

Formula Weight 156.23
Chemical Name or Material 1,7-Dimethylnaphthalene
Assay 97%
Molecular Formula C12H12
Boiling Point 263°C
MDL Number MFCD00060884
CAS 575-37-1
Beilstein 2039377
Quantity 500mg
Flash Point >110°C (230°F)

Applications
1,7-Dimethylnaphthalene can be used to produce 2,8-dimethyl-[1,4]naphthoquinone at temperature of 60. Also used as pharmaceutical intermediate.

Solubility
Soluble in water 11.5 mg/L @ 25°C.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.