Please login to your online account to display your discounted pricing

1,7-Dihydroxynaphthalene, 98%, ACROS Organics™


Quantity 5g
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  AC115010050

 View more versions of this product

Catalog No. AC115010050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder or Granules
Assay ≥97.5%
Formula Weight 160.17
Quantity 5g
Color Yellow to gray to red-brown
Beilstein 06, 981
Packaging Glass Bottle
Melting Point 178° to 182°C
CAS Max % 100.0
Synonym 1, 7-Naphthalenediol
Infrared Spectrum Authentic
Name Note 98%
Assay Percent Range 98%
Molecular Formula C10H8O2
Linear Formula C10H6(OH)2
MDL Number MFCD00035720
CAS 575-38-2
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Chemical Name or Material 1, 7-Dihydroxynaphthalene
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 1, 7-Dihydroxynaphthalene
Name Note 98%
CAS 575-38-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H8O2
Linear Formula C10H6(OH)2
Formula Weight 160.17
MDL Number MFCD00035720
Synonym 1, 7-Naphthalenediol
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents