Please login to your online account to display your discounted pricing

1,7-Dihydroxynaphthalene, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1, 7-Dihydroxynaphthalene
Name Note 98%
CAS 575-38-2
CAS Min % 97.5
CAS Max % 100.0
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C10H8O2
Linear Formula C10H6(OH)2
Formula Weight 160.17g/mol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC115010050

  SDS
thermo fisher scientific
115010050
Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass Bottle
Melting Point 178° to 182°C
Color Yellow to gray to red-brown
Quantity 5g
Chemical Name or Material 1, 7-Dihydroxynaphthalene
Name Note 98%
CAS 575-38-2
CAS Min % 97.5
CAS Max % 100.0
Physical Form Yellow to gray to red-brown powder or granules
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C10H8O2
Linear Formula C10H6(OH)2
Formula Weight 160.17g/mol
MDL Number MFCD00035720
Beilstein 06, 981
Synonym 1, 7-Naphthalenediol
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.