Learn More
1,7-Dihydroxynaphthalene, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H2826106
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
1,7-Dihydroxynaphthalene | |
180°C to 184°C | |
MFCD00035720 | |
1,7-dihydroxynaphthalene, 1,7-naphthalenediol, napthalene-1,7-diol, unii-7d42f605cs, ccris 7895, 1,7 dihydroxynaphthalene, 2,8-dihydroxynaphthalene, acmc-1aqz2, unii-m7uz7bfo3a component | |
ZUVBIBLYOCVYJU-UHFFFAOYSA-N | |
naphthalene-1,7-diol | |
68462 | |
97% |
575-38-2 | |
C10H8O2 | |
5 g | |
Slightly soluble in water. | |
C1=CC2=C(C=C(C=C2)O)C(=C1)O | |
160.172 | |
160.17 |
Chemical Identifiers
575-38-2 | |
160.172 | |
ZUVBIBLYOCVYJU-UHFFFAOYSA-N | |
68462 | |
C1=CC2=C(C=C(C=C2)O)C(=C1)O |
C10H8O2 | |
MFCD00035720 | |
1,7-dihydroxynaphthalene, 1,7-naphthalenediol, napthalene-1,7-diol, unii-7d42f605cs, ccris 7895, 1,7 dihydroxynaphthalene, 2,8-dihydroxynaphthalene, acmc-1aqz2, unii-m7uz7bfo3a component | |
naphthalene-1,7-diol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 209-383-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only