Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,7-Bis(tert-butoxycarbonylmethyl)-1,4,7,10-tetraazacyclododecane

$706.53 - $706.53

Chemical Identifiers

Chemical Name or Material 1,7-Bis(tert-butoxycarbonylmethyl)-1,4,7,10-tetraazacyclododecane
CAS 162148-48-3
Molecular Formula C20H40N4O4
Formula Weight 400.56
MDL Number MFCD09263303
Synonym Cyclen-1,7-diacetic acid di-tert-butyl ester; DO2AtBu
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH26293MD

 
alfa aesar™
H26293MD
250mg Each for $706.53
Description & Specifications

Specifications

Formula Weight 400.56
Chemical Name or Material 1,7-Bis(tert-butoxycarbonylmethyl)-1,4,7,10-tetraazacyclododecane
Molecular Formula C20H40N4O4
MDL Number MFCD09263303
CAS 162148-48-3
Synonym Cyclen-1,7-diacetic acid di-tert-butyl ester; DO2AtBu
Quantity 250mg
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Applications
Intermediate in the preparation of chelating agents.

Solubility
Soluble in acetone, acetonitrile, ethyl Acetate, chloroforme, cyclohexane. Insoluble in water.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water and moisture.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.