Please login to your online account to display your discounted pricing

1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one oxime, 97%, Maybridge


Quantity 1g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  BTB09928DA

 View more versions of this product

Catalog No. BTB09928DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 167.25
Quantity 1g
Assay 97%
CAS Max % 100.0
Chemical Name or Material 1, 7, 7-Trimethylbicyclo[2.2.1]heptan-2-one oxime
Assay Percent Range 97%
CAS 13559-66-5
Molecular Formula C10H17NO
MDL Number MFCD00210161
CAS Min % 96.5
Packaging Amber glass bottle

Specifications

Chemical Name or Material 1, 7, 7-Trimethylbicyclo[2.2.1]heptan-2-one oxime
CAS 13559-66-5
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H17NO
Formula Weight 167.25
MDL Number MFCD00210161