missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one oxime, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 13559-66-5 |
---|---|
Molecular Formula | C10H17NO |
Molecular Weight (g/mol) | 167.252 |
MDL Number | MFCD00210161 |
InChI Key | OVFDEGGJFJECAT-DHZHZOJOSA-N |
Synonym | +-camphor oxime, 1,7,7-trimethylbicyclo 2.2.1 heptan-2-one oxime, 2-bornanone oxime, 2-camphanone oxime, bornan-2-one oxime, camphor oxime, camphor, d-oxime, camphor, oxime 6ci,7ci,8ci, camphoroxime, d-camphor oxime |
PubChem CID | 5908504 |
IUPAC Name | (NE)-N-(4,7,7-trimethyl-3-bicyclo[2.2.1]heptanylidene)hydroxylamine |
SMILES | CC1(C2CCC1(C(=NO)C2)C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
BTB09928DA
|
Fisher Scientific Company
BTB09928DA |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BTB09928EA
|
Fisher Scientific Company
BTB09928EA |
10g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BTB09928EB
|
Fisher Scientific Company
BTB09928EB |
25g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
13559-66-5 | |
167.252 | |
OVFDEGGJFJECAT-DHZHZOJOSA-N | |
5908504 | |
CC1(C2CCC1(C(=NO)C2)C)C |
C10H17NO | |
MFCD00210161 | |
+-camphor oxime, 1,7,7-trimethylbicyclo 2.2.1 heptan-2-one oxime, 2-bornanone oxime, 2-camphanone oxime, bornan-2-one oxime, camphor oxime, camphor, d-oxime, camphor, oxime 6ci,7ci,8ci, camphoroxime, d-camphor oxime | |
(NE)-N-(4,7,7-trimethyl-3-bicyclo[2.2.1]heptanylidene)hydroxylamine |
Specifications
13559-66-5 | |
100.0 | |
C10H17NO | |
Amber glass bottle | |
+-camphor oxime, 1,7,7-trimethylbicyclo 2.2.1 heptan-2-one oxime, 2-bornanone oxime, 2-camphanone oxime, bornan-2-one oxime, camphor oxime, camphor, d-oxime, camphor, oxime 6ci,7ci,8ci, camphoroxime, d-camphor oxime | |
CC1(C2CCC1(C(=NO)C2)C)C | |
167.252 | |
167.25 | |
1, 7, 7-Trimethylbicyclo[2.2.1]heptan-2-one oxime |
96.5 | |
97% | |
MFCD00210161 | |
1g | |
OVFDEGGJFJECAT-DHZHZOJOSA-N | |
(NE)-N-(4,7,7-trimethyl-3-bicyclo[2.2.1]heptanylidene)hydroxylamine | |
5908504 | |
97% |