missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one oxime, 97%, Maybridge™
Manufacturer: Fisher Scientific Company BTB09928EA
Specifications
1, 7, 7-Trimethylbicyclo[2.2.1]heptan-2-one oxime | |
10g | |
96.5 | |
97% | |
MFCD00210161 | |
OVFDEGGJFJECAT-DHZHZOJOSA-N | |
(NE)-N-(4,7,7-trimethyl-3-bicyclo[2.2.1]heptanylidene)hydroxylamine | |
5908504 | |
97% |
Amber glass bottle | |
13559-66-5 | |
100.0 | |
C10H17NO | |
+-camphor oxime, 1,7,7-trimethylbicyclo 2.2.1 heptan-2-one oxime, 2-bornanone oxime, 2-camphanone oxime, bornan-2-one oxime, camphor oxime, camphor, d-oxime, camphor, oxime 6ci,7ci,8ci, camphoroxime, d-camphor oxime | |
CC1(C2CCC1(C(=NO)C2)C)C | |
167.252 | |
167.25 |
Chemical Identifiers
13559-66-5 | |
167.252 | |
OVFDEGGJFJECAT-DHZHZOJOSA-N | |
5908504 | |
CC1(C2CCC1(C(=NO)C2)C)C |
C10H17NO | |
MFCD00210161 | |
+-camphor oxime, 1,7,7-trimethylbicyclo 2.2.1 heptan-2-one oxime, 2-bornanone oxime, 2-camphanone oxime, bornan-2-one oxime, camphor oxime, camphor, d-oxime, camphor, oxime 6ci,7ci,8ci, camphoroxime, d-camphor oxime | |
(NE)-N-(4,7,7-trimethyl-3-bicyclo[2.2.1]heptanylidene)hydroxylamine |