missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(6-methylpyrazin-2-yl)piperidine-3-carbaldehyde, 97%, Maybridge

Chemical Identifiers

CAS 941716-81-0
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C11H15N3O
Formula Weight 205.26
MDL Number MFCD09879915
Chemical Name or Material 1-(6-Methylpyrazin-2-yl)piperidine-3-carbaldehyde
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC59004CB SDS Fisher Scientific Company
CC59004CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC59004DA SDS Fisher Scientific Company
CC59004DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

941716-81-0
100.0
C11H15N3O
MFCD09879915
97%
96.5
97%
205.26
1-(6-Methylpyrazin-2-yl)piperidine-3-carbaldehyde

Specifications

250mg
96.5
Amber Glass Bottle
C11H15N3O
MFCD09879915
97%
941716-81-0
100.0
97%
205.26
1-(6-Methylpyrazin-2-yl)piperidine-3-carbaldehyde

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning