missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

[1-(6-Methylpyrazin-2-yl)piperid-3-yl]methanol, 95%, Maybridge

Chemical Identifiers

CAS 937795-91-0
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C11H17N3O
Formula Weight 207.28
MDL Number MFCD09879916
Chemical Name or Material [1-(6-Methylpyrazin-2-yl)piperid-3-yl]methanol
Assay 95%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC59009CB SDS Fisher Scientific Company
CC59009CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC59009DA SDS Fisher Scientific Company
CC59009DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

937795-91-0
100.0
C11H17N3O
MFCD09879916
95%
94.5
95%
207.28
[1-(6-Methylpyrazin-2-yl)piperid-3-yl]methanol

Specifications

207.28
100.0
937795-91-0
94.5
C11H17N3O
95%
250mg
95%
MFCD09879916
Amber Glass Bottle
[1-(6-Methylpyrazin-2-yl)piperid-3-yl]methanol

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning