Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(6-Methyl-2-pyridyl)homopiperazine, 99%

Chemical Identifiers

Chemical Name or Material 1-(6-Methyl-2-pyridyl)homopiperazine
CAS 880361-95-5
Molecular Formula C11H17N3
Formula Weight 191.28
MDL Number MFCD08061041
UN Number UN3259
Synonym 1-(6-Methylpyridin-2-yl)-1,4-diazepane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5087603

  SDS
alfa aesar™
H5087603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH50876MD

  SDS
alfa aesar™
H50876MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 191.28
UN Number UN3259
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Synonym 1-(6-Methylpyridin-2-yl)-1,4-diazepane
Molecular Formula C11H17N3
MDL Number MFCD08061041
Assay 99%
Chemical Name or Material 1-(6-Methyl-2-pyridyl)homopiperazine
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Quantity 1g
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
CAS 880361-95-5
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.