Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(6-Methoxy-2-pyridyl)piperazine, 97%

$103.76 - $355.57

Chemical Identifiers

Chemical Name or Material 1-(6-Methoxy-2-pyridyl)piperazine
CAS 51047-54-2
Molecular Formula C10H15N3O
Formula Weight 193.25
MDL Number MFCD06738851
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5089503

  SDS
alfa aesar™
H5089503
1g Each for $355.57

AAH50895MD

  SDS
alfa aesar™
H50895MD
250mg Each for $103.76
Description & Specifications

Specifications

Formula Weight 193.25
Chemical Name or Material 1-(6-Methoxy-2-pyridyl)piperazine
Assay 97%
Molecular Formula C10H15N3O
MDL Number MFCD06738851
CAS 51047-54-2
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.