missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(6-Methoxy-2-naphthyl)ethan-1-one, 97%, Maybridge

Chemical Identifiers

CAS 3900-45-6
CAS Min % 97.0
Assay Percent Range 97%
Molecular Formula C13H12O2
Linear Formula CH3OC10H6COCH3
Formula Weight 200.24
Chemical Name or Material 1-(6-Methoxy-2-naphthyl)ethan-1-one
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
SB01621EA SDS Fisher Scientific Company
SB01621EA
Amber Glass Bottle 10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

3900-45-6
97%
CH3OC10H6COCH3
1-(6-Methoxy-2-naphthyl)ethan-1-one
97.0
C13H12O2
200.24

Specifications

10g
97.0
97%
CH3OC10H6COCH3
1-(6-Methoxy-2-naphthyl)ethan-1-one
3900-45-6
Amber Glass Bottle
C13H12O2
200.24

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning