missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,6-Hexanediol Dimethacrylate (stabilized with MEHQ), TCI America™

$48.89 - $184.86

Chemical Identifiers

CAS 6606-59-3
Molecular Formula C14H22O4
Molecular Weight (g/mol) 254.326
MDL Number MFCD00015425
InChI Key SAPGBCWOQLHKKZ-UHFFFAOYSA-N
Synonym 1,6-hexanediol dimethacrylate, unii-rao49190jv, 1,6-hexanediyl bismethacrylate, ccris 7049, 1,6-hexamethylene dimethacrylate, 2-propenoic acid, 2-methyl-, 1,6-hexanediyl ester, hexane-1,6-diyl bis 2-methylacrylate, 2-propenoic acid, 2-methyl-, 1,1'-1,6-hexanediyl ester, 1,6-hexanediyl bis 2-methylacrylate, 6-2-methylprop-2-enoyl oxy hexyl 2-methylprop-2-enoate
PubChem CID 81080
IUPAC Name 6-(2-methylprop-2-enoyloxy)hexyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCCCCCOC(=O)C(=C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H136925G View Documents TCI America
H136925G
25g Each for $48.89
 
H1369500G View Documents TCI America
H1369500G
500g Each for $184.86
 
Specifications

Chemical Identifiers

6606-59-3
254.326
SAPGBCWOQLHKKZ-UHFFFAOYSA-N
81080
CC(=C)C(=O)OCCCCCCOC(=O)C(=C)C
C14H22O4
MFCD00015425
1,6-hexanediol dimethacrylate, unii-rao49190jv, 1,6-hexanediyl bismethacrylate, ccris 7049, 1,6-hexamethylene dimethacrylate, 2-propenoic acid, 2-methyl-, 1,6-hexanediyl ester, hexane-1,6-diyl bis 2-methylacrylate, 2-propenoic acid, 2-methyl-, 1,1'-1,6-hexanediyl ester, 1,6-hexanediyl bis 2-methylacrylate, 6-2-methylprop-2-enoyl oxy hexyl 2-methylprop-2-enoate
6-(2-methylprop-2-enoyloxy)hexyl 2-methylprop-2-enoate

Specifications

Yellow
6606-59-3
MFCD00015425
SAPGBCWOQLHKKZ-UHFFFAOYSA-N
6-(2-methylprop-2-enoyloxy)hexyl 2-methylprop-2-enoate
81080
Clear Liquid at 20°C
1,6-Hexanediol Dimethacrylate (stabilized with MEHQ)
25g
C14H22O4
1,6-hexanediol dimethacrylate, unii-rao49190jv, 1,6-hexanediyl bismethacrylate, ccris 7049, 1,6-hexamethylene dimethacrylate, 2-propenoic acid, 2-methyl-, 1,6-hexanediyl ester, hexane-1,6-diyl bis 2-methylacrylate, 2-propenoic acid, 2-methyl-, 1,1'-1,6-hexanediyl ester, 1,6-hexanediyl bis 2-methylacrylate, 6-2-methylprop-2-enoyl oxy hexyl 2-methylprop-2-enoate
CC(=C)C(=O)OCCCCCCOC(=O)C(=C)C
254.326
254.33
≥98.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0958&(2)-1059

TSCA : Yes