Learn More
1,6-Hexanediol, 97%, Thermo Scientific Chemicals
$46.99 - $357.93
Chemical Identifiers
CAS | 629-11-8 |
---|---|
Molecular Formula | C6H14O2 |
Molecular Weight (g/mol) | 118.176 |
MDL Number | MFCD00002985 |
InChI Key | XXMIOPMDWAUFGU-UHFFFAOYSA-N |
Synonym | 1,6-hexanediol, hexamethylene glycol, 1,6-dihydroxyhexane, hexamethylenediol, alpha,omega-hexanediol, .alpha.,.omega.-hexanediol, unii-zia319275i, ccris 8982, 6-hydroxy-1-hexanol, 1,6-hexylene glycol |
PubChem CID | 12374 |
ChEBI | CHEBI:43078 |
IUPAC Name | hexane-1,6-diol |
SMILES | C(CCCO)CCO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1243930
|
Thermo Scientific Chemicals
A1243930 |
250 g |
Each for $46.99
|
|
|||||
AAA1243936
|
Thermo Scientific Chemicals
A1243936 |
500 g |
Each for $76.93
|
|
|||||
AAA124390E
|
Thermo Scientific Chemicals
A124390E |
2500 g |
Each for $357.93
|
|
|||||
Description
1,6-Hexanediol is used in polymer synthesis such as polyester, polyurethane and nylon. It is used as an intermediate to adhesives, acrylics and dyestuffs. Further, it is employed in gasoline refining and pharmaceutical production.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,6-Hexanediol is used in polymer synthesis such as polyester, polyurethane and nylon. It is used as an intermediate to adhesives, acrylics and dyestuffs. Further, it is employed in gasoline refining and pharmaceutical production.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with acid chlorides, acid anhydrides, oxidizing agents, chloroformates and reducing agents.
Chemical Identifiers
629-11-8 | |
118.176 | |
XXMIOPMDWAUFGU-UHFFFAOYSA-N | |
12374 | |
hexane-1,6-diol |
C6H14O2 | |
MFCD00002985 | |
1,6-hexanediol, hexamethylene glycol, 1,6-dihydroxyhexane, hexamethylenediol, alpha,omega-hexanediol, .alpha.,.omega.-hexanediol, unii-zia319275i, ccris 8982, 6-hydroxy-1-hexanol, 1,6-hexylene glycol | |
CHEBI:43078 | |
C(CCCO)CCO |
Specifications
629-11-8 | |
1.12 | |
147°C (296°F) | |
C6H14O2 | |
250 g | |
Hygroscopic | |
1,6-hexanediol, hexamethylene glycol, 1,6-dihydroxyhexane, hexamethylenediol, alpha,omega-hexanediol, .alpha.,.omega.-hexanediol, unii-zia319275i, ccris 8982, 6-hydroxy-1-hexanol, 1,6-hexylene glycol | |
XXMIOPMDWAUFGU-UHFFFAOYSA-N | |
hexane-1,6-diol | |
12374 | |
118.18 | |
1,6-Hexanediol |
40°C to 43°C | |
248°C to 250°C | |
97% | |
MFCD00002985 | |
1633461 | |
14,4690 | |
Soluble in water. | |
C(CCCO)CCO | |
118.176 | |
CHEBI:43078 | |
97% |
Safety and Handling
H500
EINECSNumber : 211-074-0
RTECSNumber : MO2100000
TSCA : Yes
storageNote1 : Hygroscopic
Recommended Storage : Ambient temperatures
RUO – Research Use Only