missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,6-Hexanediol, 97%

Manufacturer:  Alfa Aesar™ A1243930

 View more versions of this product

Catalog No. AAA1243930


 

Description

Description

Applications
1,6-Hexanediol is used in polymer synthesis such as polyester, polyurethane and nylon. It is used as an intermediate to adhesives, acrylics and dyestuffs. Further, it is employed in gasoline refining and pharmaceutical production.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with acid chlorides, acid anhydrides, oxidizing agents, chloroformates and reducing agents.
Specifications

Specifications

1,6-Hexanediol
248°C to 250°C
250g
97%
C6H14O2
1633461
Hexamethylene glycol
C(CCCO)CCO
118.176
CHEBI:43078
147°C (296°F)
40°C to 43°C
629-11-8
97%
MFCD00002985
14,4690
XXMIOPMDWAUFGU-UHFFFAOYSA-N
hexane-1,6-diol
12374
118.18

Chemical Identifiers

629-11-8
118.176
XXMIOPMDWAUFGU-UHFFFAOYSA-N
12374
C6H14O2
MFCD00002985
Hexamethylene glycol
CHEBI:43078
Safety and Handling

Safety and Handling

RTECSNumber : MO2100000

TSCA : TSCA

storageNote1 : Hygroscopic

SDS Documents