Please login to your online account to display your discounted pricing

1,6-Hexanediamine Dihydrochloride 99%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  411571000

 View more versions of this product

Catalog No. AC411571000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 189.13
Physical Form Crystalline Powder
Quantity 100g
Color Almost white
Merck Index 15, 4727
TSCA TSCA
EINECS Number 227-977-8
RTECS Number MO1312000
Melting Point 255° to 258°C
CAS Max % 100.0
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 98.5
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Infrared Spectrum Authentic
Health Hazard 1 HYGROSCOPIC
Molecular Formula C6H16N2·2HCl
Chemical Name or Material 1, 6-Hexanediamine dihydrochloride
MDL Number MFCD00012528
CAS 6055-52-3
Name Note 99%
Assay Percent Range 99%
Linear Formula NH2(CH2)6NH2·2HCl

Specifications

Chemical Name or Material 1, 6-Hexanediamine dihydrochloride
Name Note 99%
CAS 6055-52-3
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H16N2·2HCl
Linear Formula NH2(CH2)6NH2·2HCl
Formula Weight 189.13
MDL Number MFCD00012528
Accessories

HYGROSCOPIC

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.