missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,6-Hexanediamine Dihydrochloride 99%, ACROS Organics™

$186.95 - $186.95

Chemical Identifiers

CAS 6055-52-3
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C6H16N2·2HCl
Linear Formula NH2(CH2)6NH2·2HCl
Formula Weight 189.13
MDL Number MFCD00012528
Chemical Name or Material 1, 6-Hexanediamine dihydrochloride, 99%
Assay ≥98.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC411571000 Thermo Fisher Scientific
411571000
Glass bottle 100g Each for $186.95
Specifications Description & Specifications

Chemical Identifiers

6055-52-3
100.0
C6H16N2·2HCl
189.13
1, 6-Hexanediamine dihydrochloride, 99%
98.5
99%
NH2(CH2)6NH2·2HCl
MFCD00012528
≥98.5%

Specifications

Almost white
100g
98.5
Authentic
Crystalline Powder
C6H16N2·2HCl
189.13
15, 4727
≥98.5%
255° to 258°C
6055-52-3
100.0
Glass bottle
99%
NH2(CH2)6NH2·2HCl
MFCD00012528
1, 6-Hexanediamine dihydrochloride, 99%

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

HYGROSCOPIC

EINECSNumber : 227-977-8

RTECSNumber : MO1312000

TSCA : TSCA