missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,6-Hexanediamine, 99.5+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 120645000

 View more versions of this product

Catalog No. AC120645000



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1, 6-Hexanediamine
38.0°C to 41.0°C
204.0°C to 205.0°C
Authentic
Plastic bottle
H2N(CH2)6NH2
04,269
1,6-hexanediamine, 1,6-diaminohexane, hexamethylenediamine, hmda, 1,6-hexylenediamine, 1,6-hexamethylenediamine, 1,6-diamino-n-hexane, hexamethylene diamine, hexylenediamine, 1,6-hexanediamine solution
NAQMVNRVTILPCV-UHFFFAOYSA-N
hexane-1,6-diamine
16402
116.21
Crystalline Low Melting Solid
124-09-4
White
81°C
99.5% min. (GC)
C6H16N2
500 g
15,4727
Solubility in water: 490g/L (20°C). Other solubilities: soluble in ethanol, benzene, diethyl ether
C(CCCN)CCN
116.21
CHEBI:39618
99.5+%

Chemical Identifiers

124-09-4
116.21
1,6-hexanediamine, 1,6-diaminohexane, hexamethylenediamine, hmda, 1,6-hexylenediamine, 1,6-hexamethylenediamine, 1,6-diamino-n-hexane, hexamethylene diamine, hexylenediamine, 1,6-hexanediamine solution
CHEBI:39618
C(CCCN)CCN
C6H16N2
NAQMVNRVTILPCV-UHFFFAOYSA-N
16402
hexane-1,6-diamine
Safety and Handling

Safety and Handling

GHS H Statement
Harmful in contact with skin.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with wate

GHS Signal Word: Danger

EINECSNumber : 204-679-6

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only