Learn More
1,6-Hexanediamine, 99.5+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 120641000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1, 6-Hexanediamine | |
38.0°C to 41.0°C | |
204.0°C to 205.0°C | |
Authentic | |
Plastic bottle | |
H2N(CH2)6NH2 | |
04,269 | |
1,6-hexanediamine, 1,6-diaminohexane, hexamethylenediamine, hmda, 1,6-hexylenediamine, 1,6-hexamethylenediamine, 1,6-diamino-n-hexane, hexamethylene diamine, hexylenediamine, 1,6-hexanediamine solution | |
NAQMVNRVTILPCV-UHFFFAOYSA-N | |
hexane-1,6-diamine | |
16402 | |
116.21 | |
Crystalline Low Melting Solid |
124-09-4 | |
White | |
81°C | |
99.5% min. (GC) | |
C6H16N2 | |
100 g | |
15,4727 | |
Solubility in water: 490g/L (20°C). Other solubilities: soluble in ethanol, benzene, diethyl ether | |
C(CCCN)CCN | |
116.21 | |
CHEBI:39618 | |
99.5+% |
Chemical Identifiers
124-09-4 | |
116.21 | |
1,6-hexanediamine, 1,6-diaminohexane, hexamethylenediamine, hmda, 1,6-hexylenediamine, 1,6-hexamethylenediamine, 1,6-diamino-n-hexane, hexamethylene diamine, hexylenediamine, 1,6-hexanediamine solution | |
CHEBI:39618 | |
C(CCCN)CCN |
C6H16N2 | |
NAQMVNRVTILPCV-UHFFFAOYSA-N | |
16402 | |
hexane-1,6-diamine |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with wate
GHS Signal Word: Danger
EINECSNumber : 204-679-6
RUO – Research Use Only