Please login to your online account to display your discounted pricing

1,6-Heptadiyne, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC309420010

 View more versions of this product

Catalog No. AC309420010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 92.15
Quantity 1g
Color Clear yellow
EINECS Number 219-253-5
TSCA TSCA
Density 0.8050g/mL
RTECS Number MI5600000
CAS Max % 100.0
Packaging Glass bottle
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Highly flammable liquid and vapor.
Chemical Name or Material 1, 6-Heptadiyne
Infrared Spectrum Authentic
Refractive Index 1.4400 to 1.4440 (20°C, 589nm)
MDL Number MFCD00014925
CAS 2396-63-6
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 98%
Assay Percent Range 98%
Molecular Formula C7H8
Linear Formula CHidentical toC(CH2)3Cidentical toCH

Specifications

Chemical Name or Material 1, 6-Heptadiyne
Name Note 98%
CAS 2396-63-6
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H8
Linear Formula CHidentical toC(CH2)3Cidentical toCH
Formula Weight 92.15
MDL Number MFCD00014925
Accessories

GHS Signal Word: Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Highly flammable liquid and vapor.

SDS & Safety Documents