missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,6-Heptadiyne, 97%

Manufacturer:  Alfa Aesar™ A1131806

 View more versions of this product

Catalog No. AAA1131806



Description

Description

Applications
1,6-Heptadiyne may be used in the preparation of free-standing polymer films of poly(1,6-heptadiyne). Polymerization of 1,6-heptadiyne using an insoluble Zeigler-Natta catalyst affords soluble polymer having six-membered ring containing polyene. Ruthenium(II)-catalyzed reaction of a substituted 1,6-heptadiyne with norbornene affords a tandem [2+ 2+ 2]/[4+2] cycloaddition product and a [2+ 2+2] cycloadduct.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Haevy metal powder, heat. Refrigerate at (+4°C).
Specifications

Specifications

1,6-Heptadiyne
Recognizable
5g
C7H8
UN3295
1,6-heptadiyne, 1,6-heptadiyne 6ci, 8ci, 9ci, heptane-1,6-diyne, 1,6-heptadiyne, 1,6-heptadiyne, acmc-1cin6, 1,6-heptadiyne, 1,6-heptadiyne, 97+%, wln: 1uu5uu1, 4-01-00-01121 beilstein handbook reference
C#CCCCC#C
92.141
92.14
9°C (48°F)
109°C to 111°C
2396-63-6
MFCD00014925
1731756
RSPZSDWVQWRAEF-UHFFFAOYSA-N
hepta-1,6-diyne
337121
97%

Chemical Identifiers

2396-63-6
92.141
RSPZSDWVQWRAEF-UHFFFAOYSA-N
337121
C#CCCCC#C
C7H8
MFCD00014925
1,6-heptadiyne, 1,6-heptadiyne 6ci, 8ci, 9ci, heptane-1,6-diyne, 1,6-heptadiyne, 1,6-heptadiyne, acmc-1cin6, 1,6-heptadiyne, 1,6-heptadiyne, 97+%, wln: 1uu5uu1, 4-01-00-01121 beilstein handbook reference
hepta-1,6-diyne
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.

GHS P Statement
P210-P243-P301+P330+P331-P315
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take precautionary measures against static discharge.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Get immediate medical advice/attention.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

RTECSNumber : MI5600000

TSCA : TSCA

Recommended Storage : 2°C to 8°C

SDS Documents