Learn More
1,6-Heptadien-4-ol, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 150090050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1, 6-Heptadien-4-ol | |
2883-45-6 | |
100.0 | |
0.8600g/mL | |
40°C | |
96% min. (GC) | |
C7H12O | |
H2C=CHCH2CH(OH)CH2CH=CH2 | |
5 g | |
0.86 | |
UTGFOWQYZKTZTN-UHFFFAOYSA-N | |
hepta-1,6-dien-4-ol | |
17902 | |
97% |
97% | |
96.0 | |
Colorless to Yellow | |
151°C | |
Authentic | |
Glass bottle | |
1.4490 to 1.453 | |
MFCD00008663 | |
01, 455 | |
1,6-heptadien-4-ol, 1,6-heptadiene-4-ol, diallylcarbinol, pubchem13740, 4-hepta-1,6-dienol, acmc-1cei5, intermediates-zcf02285, 4-hydroxy-1,6-heptadiene, 4-01-00-02249 beilstein handbook reference | |
OC(CC=C)CC=C | |
112.17 | |
112.17 | |
Liquid |
Chemical Identifiers
2883-45-6 | |
112.17 | |
UTGFOWQYZKTZTN-UHFFFAOYSA-N | |
17902 | |
OC(CC=C)CC=C |
C7H12O | |
MFCD00008663 | |
1,6-heptadien-4-ol, 1,6-heptadiene-4-ol, diallylcarbinol, pubchem13740, 4-hepta-1,6-dienol, acmc-1cei5, intermediates-zcf02285, 4-hydroxy-1,6-heptadiene, 4-01-00-02249 beilstein handbook reference | |
hepta-1,6-dien-4-ol |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wash face,hands and any exposed skin thoroughly
GHS Signal Word: Warning
EINECSNumber : 220-742-0
RTECSNumber : MI5450000
TSCA : TSCA
RUO – Research Use Only