missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(6-Ethoxy-2-pyridyl)piperazine, 97%

$99.18 - $342.57

Chemical Identifiers

CAS 108122-25-4
Molecular Formula C11H17N3O
Formula Weight 207.28
MDL Number MFCD08061051
Synonym 2-Ethoxy-6-(1-piperazinyl)pyridine
Chemical Name or Material 1-(6-Ethoxy-2-pyridyl)piperazine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5090503 SDS Alfa Aesar™
H5090503
1g Each for $342.57
AAH50905MD SDS Alfa Aesar™
H50905MD
250mg Each for $99.18
Specifications Description & Specifications

Chemical Identifiers

108122-25-4
207.28
2-Ethoxy-6-(1-piperazinyl)pyridine
C11H17N3O
MFCD08061051
1-(6-Ethoxy-2-pyridyl)piperazine

Specifications

1g
C11H17N3O
MFCD08061051
1-(6-Ethoxy-2-pyridyl)piperazine
108122-25-4
207.28
2-Ethoxy-6-(1-piperazinyl)pyridine
97%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning