Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,6-Diphenoxy-2,4-hexadiyne, 98+%


Quantity 5g

Manufacturer: alfa aesar™  L1000406

 View more versions of this product

Catalog No. AAL1000406

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 262.31
Chemical Name or Material 1,6-Diphenoxy-2,4-hexadiyne
Assay ≥98%
Molecular Formula C18H14O2
MDL Number MFCD00041712
Linear Formula C6H5OCH2C≡C-C≡CCH2OC6H5
CAS 30980-37-1
Beilstein 1886340
Quantity 5g
Melting Point 80° to 83°C

Specifications

Chemical Name or Material 1,6-Diphenoxy-2,4-hexadiyne
CAS 30980-37-1
Molecular Formula C18H14O2
Linear Formula C6H5OCH2C≡C-C≡CCH2OC6H5
Formula Weight 262.31
MDL Number MFCD00041712
Beilstein 1886340