Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,6-Diphenoxy-2,4-hexadiyne, 98+%

$85.27 - $342.04

Chemical Identifiers

Chemical Name or Material 1,6-Diphenoxy-2,4-hexadiyne
CAS 30980-37-1
Molecular Formula C18H14O2
Linear Formula C6H5OCH2C≡C-C≡CCH2OC6H5
Formula Weight 262.31
MDL Number MFCD00041712
Beilstein 1886340
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1000406

  SDS
alfa aesar™
L1000406
5g Each for $85.27

AAL1000414

  SDS
alfa aesar™
L1000414
25g Each for $342.04
Description & Specifications

Specifications

Formula Weight 262.31
MDL Number MFCD00041712
Molecular Formula C18H14O2
Chemical Name or Material 1,6-Diphenoxy-2,4-hexadiyne
Assay ≥98%
CAS 30980-37-1
Quantity 5g
Beilstein 1886340
Linear Formula C6H5OCH2C≡C-C≡CCH2OC6H5
Melting Point 80° to 83°C