Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,6-Dimethoxyhexane, 98%

$37.36 - $357.11

Chemical Identifiers

Chemical Name or Material 1,6-Dimethoxyhexane
CAS 13179-98-1
Molecular Formula CH3O(CH2)6OCH3
Formula Weight 146.23
MDL Number MFCD00142569
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2513006

  SDS
alfa aesar™
B2513006
5g Each for $37.36

AAB2513022

  SDS
alfa aesar™
B2513022
100g Each for $357.11
Description & Specifications

Specifications

Formula Weight 146.23
Chemical Name or Material 1,6-Dimethoxyhexane
Assay 98%
Molecular Formula CH3O(CH2)6OCH3
Boiling Point 182° to 183°C
MDL Number MFCD00142569
CAS 13179-98-1
Quantity 5g
Flash Point 69°C (157°F)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.