missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,6-Diisocyanatohexane, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 04380330

 View more versions of this product

Catalog No. AA4380330



Description

Description

1,6-Diisocyanatohexane is used as a polymerizing agent in polyurethane paints and coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
Specifications

Specifications

1,6-Diisocyanatohexane
-67°C
255°C
Pungent
1.452
250 g
956709
hexamethylene diisocyanate, 1,6-hexamethylene diisocyanate, hmdi, hexane, 1,6-diisocyanato, hexane 1,6-diisocyanate, 1,6-hexylene diisocyanate, hexamethylene-1,6-diisocyanate, 1,6-hexanediol diisocyanate, hdi, isocyanic acid, hexamethylene ester
RRAMGCGOFNQTLD-UHFFFAOYSA-N
1,6-diisocyanatohexane
13192
168.19
Liquid
822-06-0
1.05 g/mL
130°C (266°F)
C8H12N2O2
MFCD00002047
UN2281
Moisture sensitive
Reacts with water.
O=C=NCCCCCCN=C=O
168.20
CHEBI:53578
≥98%

Chemical Identifiers

822-06-0
168.20
RRAMGCGOFNQTLD-UHFFFAOYSA-N
13192
1,6-diisocyanatohexane
C8H12N2O2
MFCD00002047
hexamethylene diisocyanate, 1,6-hexamethylene diisocyanate, hmdi, hexane, 1,6-diisocyanato, hexane 1,6-diisocyanate, 1,6-hexylene diisocyanate, hexamethylene-1,6-diisocyanate, 1,6-hexanediol diisocyanate, hdi, isocyanic acid, hexamethylene ester
CHEBI:53578
O=C=NCCCCCCN=C=O
Safety and Handling

Safety and Handling

GHS H Statement
H331-H334-H315-H319-H317-H335-H204
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Fire or projection hazard.

P260-P264b-P270-P271-P272-P280-P284-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c

H302-H314-H317-H330-H334-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: HEXAMETHYLENE DIISOCYANATE

EINECSNumber : 212-485-8

RTECSNumber : MO1740000

TSCA : Yes

storageNote1 :

  • Packaged under Argon

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only