Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,6-Diiodoperfluorohexane, 97%

$734.24 - $734.24

Chemical Identifiers

Chemical Name or Material 1,6-Diiodoperfluorohexane
CAS 375-80-4
Molecular Formula I(CF2)6I
Formula Weight 553.86
MDL Number MFCD00042264
Beilstein 1822186
Synonym 1,6-Diiodododecafluorohexane; Dodecafluoro-1,6-diiodohexane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2165414

  SDS
alfa aesar™
B2165414
25g Each for $734.24
Description & Specifications

Specifications

Formula Weight 553.86
Chemical Name or Material 1,6-Diiodoperfluorohexane
Melting Point 25°C
Assay 97%
Boiling Point 115°C (100mmHg)
Molecular Formula I(CF2)6I
Quantity 25g
MDL Number MFCD00042264
CAS 375-80-4
Beilstein 1822186
Synonym 1,6-Diiodododecafluorohexane; Dodecafluoro-1,6-diiodohexane

Applications
It is employed in the synthesis and characterization of fluorinated telomers containing vinylidene fluoride and hexafluoropropene.

Notes
Light sensitive. Protect from light. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.