missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,6-Dihydroxynaphthalene, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC269910250

 View more versions of this product

Catalog No. AC269910250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1, 6-Dihydroxynaphthalene
Glass bottle
138.0°C to 140.0°C
575-44-0
100.0
C10H8O2
FZZQNEVOYIYFPF-UHFFFAOYSA-N
naphthalene-1,6-diol
68463
160.17
99%
Authentic
White to Brown
25g
98.5
98.5% min. (HPLC)
1,6-dihydroxy naphthalene, 1,6-dihydroxynaphtalene, 1,6-dihydroxynaphthalene, 1,6-naphthalenediol, 1.6-dihydroxynaphthalene, 2,5-dihydroxynaphthalene, 2,5-naphthalenediol, 6-hydroxy-1-naphthol, ccris 7894, naphthalene, 1,6-dihydroxy
C1=CC2=C(C=CC(=C2)O)C(=C1)O
160.17
CHEBI:42040
Powder or Crystalline Powder

Chemical Identifiers

575-44-0
160.17
1,6-dihydroxy naphthalene, 1,6-dihydroxynaphtalene, 1,6-dihydroxynaphthalene, 1,6-naphthalenediol, 1.6-dihydroxynaphthalene, 2,5-dihydroxynaphthalene, 2,5-naphthalenediol, 6-hydroxy-1-naphthol, ccris 7894, naphthalene, 1,6-dihydroxy
CHEBI:42040
C1=CC2=C(C=CC(=C2)O)C(=C1)O
C10H8O2
FZZQNEVOYIYFPF-UHFFFAOYSA-N
68463
naphthalene-1,6-diol
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 209-386-7

SDS Documents