Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,6-Dicyanohexane, 99%

Chemical Identifiers

Chemical Name or Material 1,6-Dicyanohexane
CAS 629-40-3
Molecular Formula NC(CH2)6CN
Formula Weight 136.2
MDL Number MFCD00001981
UN Number UN3276
Beilstein 1744365
Synonym Octanedinitrile; Suberonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2454314

  SDS
alfa aesar™
B2454314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2454322

  SDS
alfa aesar™
B2454322
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 136.2
CAS 629-40-3
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Quantity 25g
Beilstein 1744365
Synonym Octanedinitrile; Suberonitrile
Assay 99%
MDL Number MFCD00001981
UN Number UN3276
TSCA TSCA
Boiling Point 174° to 176°C
Health Hazard 1 Danger
Melting Point −3°C
Chemical Name or Material 1,6-Dicyanohexane
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
Flash Point 175°C (347°F)
RTECS Number WM2113000
Molecular Formula NC(CH2)6CN
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.