Please login to your online account to display your discounted pricing

1,6-Dichlorohexane, 98%, ACROS Organics™

$37.51 - $37.51

Chemical Identifiers

Chemical Name or Material 1, 6-Dichlorohexane
CAS 2163-00-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H12Cl2
Linear Formula Cl(CH2)6Cl
Formula Weight 155.07
MDL Number MFCD00001020
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC113411000

  SDS
thermo fisher scientific
113411000
Glass bottle 100g Each for $37.51
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥97.5%
Formula Weight 155.07
Quantity 100g
Color Clear colorless
Density 1.0700g/mL
Beilstein 01, 144
CAS Max % 100.0
Chemical Name or Material 1, 6-Dichlorohexane
Linear Formula Cl(CH2)6Cl
Packaging Glass bottle
Refractive Index 1.4558 to 1.4578 (20°C, 589nm)
Molecular Formula C6H12Cl2
MDL Number MFCD00001020
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Warning
Assay Percent Range 98%
CAS 2163-00-0
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Harmful to aquatic life with long lasting effects.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Harmful to aquatic life with long lasting effects.
Causes serious eye irritation.
May cause respiratory irritation.