Please login to your online account to display your discounted pricing

1,6-Dichlorohexane, 98%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  113411000

 View more versions of this product

Catalog No. AC113411000

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 155.07
Assay ≥97.5%
Physical Form Liquid
Quantity 100g
Color Clear colorless
Density 1.0700g/mL
Beilstein 01, 144
CAS Max % 100.0
Linear Formula Cl(CH2)6Cl
Chemical Name or Material 1, 6-Dichlorohexane
Packaging Glass bottle
Molecular Formula C6H12Cl2
Health Hazard 1 GHS Signal Word: Warning
CAS Min % 97.5
MDL Number MFCD00001020
Refractive Index 1.4558 to 1.4578 (20°C, 589nm)
Infrared Spectrum Authentic
CAS 2163-00-0
Assay Percent Range 98%
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Harmful to aquatic life with long lasting effects.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 1, 6-Dichlorohexane
CAS 2163-00-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H12Cl2
Linear Formula Cl(CH2)6Cl
Formula Weight 155.07
MDL Number MFCD00001020
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Harmful to aquatic life with long lasting effects.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents