Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,6-Dibromoperfluorohexane, 96%

$50.78 - $174.78

Chemical Identifiers

Chemical Name or Material 1,6-Dibromoperfluorohexane
CAS 918-22-9
Molecular Formula Br(CF2)6Br
Formula Weight 459.87
MDL Number MFCD00153110
Beilstein 1822184
Synonym 1,6-Dibromodecafluorohexane; Perfluoro-1,6-dibromohexane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2272903

  SDS
alfa aesar™
B2272903
1g Each for $50.78

AAB2272906

  SDS
alfa aesar™
B2272906
5g Each for $174.78
Description & Specifications

Specifications

Formula Weight 459.87
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 96%
Chemical Name or Material 1,6-Dibromoperfluorohexane
Boiling Point 141° to 142°C
Beilstein 1822184
Synonym 1,6-Dibromodecafluorohexane; Perfluoro-1,6-dibromohexane
Quantity 1g
CAS 918-22-9
Health Hazard 1 Warning
MDL Number MFCD00153110
Molecular Formula Br(CF2)6Br

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.