missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,6-Dibromo-2-naphthol, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 112851000

 View more versions of this product

Catalog No. AC112851000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

1, 6-Dibromo-2-naphthol
Glass bottle
105.0°C to 107.0°C
99%
98.5
98.5% min. (GC)
Br2C10H5OH
06, 652
VKESFYLPKHQOOA-UHFFFAOYSA-N
1,6-dibromonaphthalen-2-ol
27768
Powder
Authentic
Brown or Pink to Purple
100g
16239-18-2
100.0
C10H6Br2O
MFCD00003870
1,6 dibromo-2-napthol, 1,6-dibromo-.beta.-naphthol, 1,6-dibromo-2-hydroxynaphthalene, 1,6-dibromo-2-naphthol, 1,6-dibromo-beta-naphthol, 2-naphthalenol, 1,6-dibromo, 2-naphthol, 1,6-dibromo, 2-naphthol,6-dibromo, acmc-1c8jh, pubchem22920
C1=CC2=C(C=CC(=C2Br)O)C=C1Br
301.96
301.96
99%

Chemical Identifiers

16239-18-2
301.96
VKESFYLPKHQOOA-UHFFFAOYSA-N
27768
C1=CC2=C(C=CC(=C2Br)O)C=C1Br
C10H6Br2O
MFCD00003870
1,6 dibromo-2-napthol, 1,6-dibromo-.beta.-naphthol, 1,6-dibromo-2-hydroxynaphthalene, 1,6-dibromo-2-naphthol, 1,6-dibromo-beta-naphthol, 2-naphthalenol, 1,6-dibromo, 2-naphthol, 1,6-dibromo, 2-naphthol,6-dibromo, acmc-1c8jh, pubchem22920
1,6-dibromonaphthalen-2-ol
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 240-356-6

RTECSNumber : QL3500000

TSCA : TSCA

SDS Documents