Please login to your online account to display your discounted pricing

1,6-Dibromo-2-naphthol 99%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  112850050

 View more versions of this product

Catalog No. AC112850050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Assay ≥98.5%
Formula Weight 301.96
Quantity 5g
Color Pink to purple or light brown
EINECS Number 240-356-6
TSCA TSCA
RTECS Number QL3500000
Melting Point 105° to 109°C
Beilstein 06, 652
CAS Max % 100.0
Infrared Spectrum Authentic
MDL Number MFCD00003870
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Chemical Name or Material 1, 6-Dibromo-2-naphthol
Name Note 99%
Assay Percent Range 99%
Linear Formula Br2C10H5OH
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C10H6Br2O
CAS 16239-18-2

Specifications

Chemical Name or Material 1, 6-Dibromo-2-naphthol
Name Note 99%
CAS 16239-18-2
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H6Br2O
Linear Formula Br2C10H5OH
Formula Weight 301.96
MDL Number MFCD00003870
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents