Please login to your online account to display your discounted pricing

1,6-Dibromo-2-naphthol 99%, ACROS Organics™

$82.71 - $82.71

Chemical Identifiers

Chemical Name or Material 1, 6-Dibromo-2-naphthol
Name Note 99%
CAS 16239-18-2
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H6Br2O
Linear Formula Br2C10H5OH
Formula Weight 301.96
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC112850250

  SDS
thermo fisher scientific
112850250
Glass bottle 25g Each for $82.71

AC112850050

  SDS
thermo fisher scientific
112850050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC112851000

  SDS
thermo fisher scientific
112851000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Assay ≥98.5%
Formula Weight 301.96
Quantity 25g
Color Pink to purple or light brown
Beilstein 06, 652
EINECS Number 240-356-6
TSCA TSCA
RTECS Number QL3500000
Melting Point 105° to 109°C
CAS Max % 100.0
MDL Number MFCD00003870
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 99%
Linear Formula Br2C10H5OH
CAS 16239-18-2
Infrared Spectrum Authentic
CAS Min % 98.5
Assay Percent Range 99%
Molecular Formula C10H6Br2O
Chemical Name or Material 1, 6-Dibromo-2-naphthol
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.